Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133882.2 | NP_598643.1 | 3 - 4 | 467 | 70 | |
GenBank mRNA | AB077304.1 | - | 3 - 4 | 389 | 70 | |
AB077305.1 | - | 3 - 4 | 389 | 70 | ||
AB077306.1 | - | 3 - 4 | 389 | 70 | ||
AK050313.1 | - | 3 - 4 | 468 | 70 | ||
BC022129.1 | - | 1 - 2 | 98 | 70 | ||
BC096382.1 | - | 3 - 4 | 441 | 5102825 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |