Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163103.1 | NP_001156575.1 | 4 - 5 | 370 | 83 | |
GenBank mRNA | AK133227.1 | - | 4 - 5 | 349 | 83 | |
CK137158.1 | - | 4 - 5 | 370 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |