Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3yl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013709.5 | NP_038737.1 | 2 - 3 | 671 | 119 | |
NR_110343.1 | 3 - 4 | 816 | 119 | |||
GenBank mRNA | AK141199.1 | - | 2 - 3 | 672 | 119 | |
AK168483.1 | - | 3 - 4 | 299 | 119 | ||
BC010296.1 | - | 1 - 2 | 119 | 3154966 | 119 | |
BC061488.1 | - | 2 - 3 | 611 | 4483634 | 119 | |
BC110043.1 | - | 2 - 3 | 649 | 5149633 | 119 | |
D85926.1 | - | 2 - 3 | 154 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |