Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039878.2 | NP_001034967.1 | 11 - 12 | 1938 | 63 | |
NM_133789.3 | NP_598550.2 | 11 - 12 | 1959 | 63 | ||
GenBank mRNA | AF414080.1 | - | 11 - 12 | 1959 | 63 | |
AK136155.1 | - | 11 - 12 | 1255 | 63 | ||
AK154615.1 | - | 11 - 12 | 1488 | 63 | ||
AK161673.1 | - | 11 - 12 | 1509 | 63 | ||
BC004025.1 | - | 9 - 10 | 1072 | 63 | ||
BC080283.1 | - | 11 - 12 | 1525 | 6401782 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |