Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145920.2 | NP_001139392.1 | 3 - 4 | 1678 | 107 | |
NM_001146038.2 | NP_001139510.1 | 5 - 6 | 943 | 107 | ||
NM_001271627.1 | NP_001258556.1 | 4 - 5 | 1107 | 107 | ||
NM_009820.5 | NP_033950.2 | 5 - 6 | 910 | 107 | ||
NR_073392.1 | 4 - 5 | 694 | 107 | |||
GenBank mRNA | AF005936.1 | - | 4 - 5 | 1061 | 107 | |
AF010284.1 | - | 5 - 6 | 910 | 107 | ||
AF053954.1 | - | 4 - 5 | 524 | 107 | ||
D14636.1 | - | 3 - 4 | 1676 | 107 | ||
D14637.1 | - | 3 - 4 | 1676 | 107 | ||
DQ458792.1 | - | 4 - 5 | 695 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |