Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081335.2 | NP_001074804.2 | 25 - 26 | 5184 | 88 | |
GenBank mRNA | AK051474.1 | - | 2 - 3 | 412 | 88 | |
AK122351.1 | - | 22 - 23 | 4175 | 88 | ||
AK147675.1 | - | 23 - 24 | 5774 | 88 | ||
AK162968.1 | - | 4 - 5 | 644 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |