Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008434.2 | NP_032460.2 | 4 - 5 | 787 | 75 | |
GenBank mRNA | AB079603.1 | - | 4 - 5 | 490 | 75 | |
AK154244.1 | - | 4 - 5 | 761 | 75 | ||
AY331142.1 | - | 4 - 5 | 702 | 75 | ||
BC045142.1 | - | 4 - 5 | 787 | 6314642 | 75 | |
BC055304.1 | - | 4 - 5 | 742 | 6397978 | 75 | |
U70068.1 | - | 4 - 5 | 593 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |