Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033270.2 | NP_150373.2 | 3 - 4 | 615 | 75 | |
NR_027677.1 | 4 - 5 | 681 | 75 | |||
GenBank mRNA | AB221552.1 | - | 3 - 4 | 376 | 75 | |
AK035116.1 | - | 3 - 4 | 612 | 75 | ||
AK146792.1 | - | 4 - 5 | 677 | 75 | ||
AK149061.1 | - | 771 | 75 | |||
BC019166.1 | - | 4 - 5 | 677 | 4236246 | 75 | |
BC037656.1 | - | 4 - 5 | 558 | 5358554 | 75 | |
BC057929.1 | - | 3 - 4 | 603 | 5289475 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |