Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130412.1 | NP_001123884.1 | 15 - 16 | 2109 | 80 | |
NM_015763.4 | NP_056578.2 | 15 - 16 | 2064 | 80 | ||
NM_172950.3 | NP_766538.2 | 14 - 15 | 1965 | 80 | ||
GenBank mRNA | AB093214.1 | - | 10 - 11 | 1284 | 80 | |
AF180471.1 | - | 14 - 15 | 1944 | 80 | ||
AF412811.1 | - | 14 - 15 | 2002 | 80 | ||
AK014526.1 | - | 14 - 15 | 2025 | 80 | ||
AK019539.1 | - | 15 - 16 | 2202 | 80 | ||
AK030921.1 | - | 14 - 15 | 1968 | 80 | ||
AK154837.1 | - | 15 - 16 | 2106 | 80 | ||
BC042462.1 | - | 14 - 15 | 2018 | 4211202 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |