Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134040.1 | NP_598801.1 | 23 - 24 | 1923 | 74 | |
GenBank mRNA | AK028341.1 | - | 23 - 24 | 1920 | 74 | |
AK132157.1 | - | 23 - 24 | 1963 | 74 | ||
AK153335.1 | - | 23 - 24 | 1947 | 74 | ||
AK160982.1 | - | 23 - 24 | 1924 | 74 | ||
AK166146.1 | - | 23 - 24 | 1906 | 74 | ||
BC008570.1 | - | 23 - 24 | 1901 | 3587491 | 74 | |
BC010624.1 | - | 23 - 24 | 1923 | 3153029 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |