Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027649.3 | NP_081925.2 | 2 - 3 | 297 | 118 | |
GenBank mRNA | AK005865.1 | - | 2 - 3 | 118 | ||
AK016586.1 | - | 2 - 3 | 224 | 118 | ||
AY336501.1 | - | 2 - 3 | 185 | 118 | ||
BC131909.1 | - | 2 - 3 | 257 | 40130186 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |