Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025371.2 | NP_001020542.1 | 3 - 4 | 655 | 58 | |
GenBank mRNA | AK044479.1 | - | 3 - 4 | 683 | 58 | |
BC026459.1 | - | 3 - 4 | 584 | 58 | ||
BC083132.1 | - | 3 - 4 | 655 | 6466398 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |