Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007998.mm.1 |
NM_001110145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007998.mm.1 |
NM_001110145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007998.mm.1 |
NM_009278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007998.mm.1 |
NM_009278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007998.mm.1 |
TC0200001013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007999.mm.1 |
NM_001110145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007999.mm.1 |
NM_001110145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007999.mm.1 |
NM_009278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007999.mm.1 |
NM_009278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007999.mm.1 |
TC0200001013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008002.mm.1 |
NM_001110145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008002.mm.1 |
NM_001110145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008002.mm.1 |
NM_009278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008002.mm.1 |
NM_009278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008002.mm.1 |
TC0200001013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008004.mm.1 |
NM_001110145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008004.mm.1 |
NM_001110145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008004.mm.1 |
NM_009278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008004.mm.1 |
NM_009278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008004.mm.1 |
TC0200001013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008003.mm.1 |
NM_009278.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008003.mm.1 |
NM_009278.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008003.mm.1 |
TC0200001013.mm.1 |
MTA TC |