Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133700.2 | NP_598461.1 | 2 - 3 | 231 | 72 | |
GenBank mRNA | BC050781.1 | - | 2 - 3 | 231 | 6591397 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |