Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyltl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166633.2 | NP_001160105.1 | 1 - 2 | 77 | 66 | |
NM_001290773.1 | NP_001277702.1 | 1 - 2 | 77 | 66 | ||
NM_001290774.1 | NP_001277703.1 | 1 - 2 | 77 | 66 | ||
NM_001290775.1 | NP_001277704.1 | 1 - 2 | 77 | 66 | ||
NM_172670.3 | NP_766258.2 | 1 - 2 | 77 | 66 | ||
GenBank mRNA | AK081797.1 | - | 1 - 2 | 54 | 66 | |
AY742914.1 | - | 1 - 2 | 53 | 66 | ||
AY742915.1 | - | 1 - 2 | 53 | 66 | ||
CF170217.1 | - | 1 - 2 | 56 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |