Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008914.2 | NP_032940.1 | 4 - 5 | 653 | 93 | |
GenBank mRNA | AB221678.1 | - | 4 - 5 | 520 | 93 | |
AK135203.1 | - | 4 - 5 | 467 | 93 | ||
BC037668.1 | - | 4 - 5 | 476 | 93 | ||
BC066000.1 | - | 4 - 5 | 624 | 5699179 | 93 | |
M81483.1 | - | 4 - 5 | 490 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |