Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008914.2 | NP_032940.1 | 8 - 9 | 1120 | 170 | |
GenBank mRNA | AB221678.1 | - | 8 - 9 | 987 | 170 | |
AK135203.1 | - | 8 - 9 | 934 | 170 | ||
BC037668.1 | - | 8 - 9 | 943 | 170 | ||
BC066000.1 | - | 8 - 9 | 1091 | 5699179 | 170 | |
M81483.1 | - | 8 - 9 | 957 | 170 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |