Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019439.3 | NP_062312.3 | 18 - 19 | 2535 | 67 | |
GenBank mRNA | AF008649.1 | - | 18 - 19 | 2419 | 67 | |
AF120255.1 | - | 13 - 14 | 1863 | 67 | ||
AK038844.1 | - | 1575 | 67 | |||
AK156397.1 | - | 16 - 17 | 2269 | 67 | ||
BC054735.1 | - | 18 - 19 | 2535 | 6401243 | 67 | |
BC056990.1 | - | 13 - 14 | 1912 | 6821442 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |