Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023162.4 | NP_075651.1 | 1 - 2 | 281 | 71 | |
GenBank mRNA | AF230339.1 | - | 1 - 2 | 154 | 71 | |
AK003714.1 | - | 1 - 2 | 282 | 71 | ||
AK005976.1 | - | 2 - 3 | 254 | 71 | ||
AK019152.1 | - | 2 - 3 | 258 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |