Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023712.3 | NP_076201.2 | 8 - 9 | 2004 | 61 | |
NR_045537.1 | 9 - 10 | 1983 | 61 | |||
GenBank mRNA | AF212372.1 | - | 8 - 9 | 1495 | 61 | |
AK008721.1 | - | 9 - 10 | 1480 | 61 | ||
AK033907.1 | - | 6 - 7 | 2559 | 61 | ||
AK154353.1 | - | 8 - 9 | 1494 | 61 | ||
AK166931.1 | - | 8 - 9 | 1496 | 61 | ||
AK195969.1 | - | 2 - 3 | 312 | 61 | ||
BC002297.1 | - | 8 - 9 | 1023 | 61 | ||
BC026854.1 | - | 8 - 9 | 1465 | 4242049 | 61 | |
BC085491.1 | - | 8 - 9 | 1448 | 6829415 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |