Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165935.1 | NP_001159407.1 | 3 - 4 | 280 | 68 | |
NM_028844.3 | NP_083120.2 | 4 - 5 | 610 | 68 | ||
GenBank mRNA | AK005934.1 | - | 3 - 4 | 335 | 68 | |
BC100725.1 | - | 5 - 6 | 473 | 40045263 | 68 | |
BC100735.1 | - | 5 - 6 | 487 | 68 | ||
BX528932.1 | - | 2 - 3 | 140 | 68 | ||
DV646415.1 | - | 3 - 4 | 282 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |