Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050404.1 | - | 5 - 6 | 1013 | 101 | |
BC061229.1 | - | 5 - 6 | 914 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |