Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013684.3 | NP_038712.3 | 1 - 2 | 136 | 65 | |
GenBank mRNA | AK085037.1 | - | 1 - 2 | 137 | 65 | |
BC016476.1 | - | 1 - 2 | 114 | 3584341 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |