Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146211.1 | NP_001139683.1 | 2 - 3 | 133 | 141 | |
GenBank mRNA | BC038917.1 | - | 2 - 3 | 123 | 141 | |
DV046794.1 | - | 2 - 3 | 147 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |