Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054103.1 | NP_473444.1 | 4 - 5 | 789 | 70 | |
GenBank mRNA | AK014819.1 | - | 4 - 5 | 789 | 70 | |
AM056057.1 | - | 1 - 2 | 57 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |