Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026620.3 | NP_080896.2 | 4 - 5 | 564 | 142 | |
GenBank mRNA | BC048087.1 | - | 4 - 5 | 554 | 6439798 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |