Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028708.2 | NP_082984.1 | 1294 | 83 | ||
GenBank mRNA | AK018189.2 | - | 1296 | 83 | ||
AK220482.1 | - | 1274 | 83 | |||
BC119557.1 | - | 968 | 83 | |||
BC119558.1 | - | 959 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |