Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144887.4 | NP_659136.1 | 9 - 10 | 2243 | 85 | |
GenBank mRNA | AK028737.1 | - | 6 - 7 | 809 | 85 | |
AK082513.1 | - | 9 - 10 | 2168 | 85 | ||
AK145086.1 | - | 9 - 10 | 2170 | 85 | ||
AK173251.1 | - | 9 - 10 | 2198 | 85 | ||
AY894891.1 | - | 9 - 10 | 2235 | 85 | ||
BC020051.1 | - | 8 - 9 | 1213 | 3987696 | 85 | |
BC027047.1 | - | 5 - 6 | 477 | 85 | ||
BC065155.1 | - | 9 - 10 | 2113 | 5706087 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |