Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172409.2 | NP_765997.2 | 9 - 10 | 1154 | 139 | |
GenBank mRNA | CA329111.1 | - | 5 - 6 | 525 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |