Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101488.1 | NP_001094958.1 | 1 - 2 | 552 | 65 | |
GenBank mRNA | AK082731.1 | - | 1 - 2 | 554 | 65 | |
BB651613.1 | - | 1 - 2 | 547 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |