Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009844.2 | NP_033974.2 | 5 - 6 | 969 | 68 | |
GenBank mRNA | AK089835.1 | - | 5 - 6 | 968 | 68 | |
AK156125.1 | - | 5 - 6 | 969 | 68 | ||
AK163608.1 | - | 3 - 4 | 1466 | 68 | ||
AK172008.1 | - | 5 - 6 | 931 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |