Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172620.3 | NP_766208.2 | 5 - 6 | 588 | 83 | |
GenBank mRNA | AK042520.1 | - | 5 - 6 | 542 | 83 | |
AK051732.1 | - | 5 - 6 | 659 | 83 | ||
AK052523.1 | - | 5 - 6 | 589 | 83 | ||
AK214507.1 | - | 1 - 2 | 80 | 83 | ||
AX775735.1 | - | 5 - 6 | 400 | 83 | ||
KJ650214.1 | - | 5 - 6 | 377 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |