Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011280.2 | NP_035410.2 | 5 - 6 | 1200 | 63 | |
GenBank mRNA | AF134811.1 | - | 5 - 6 | 1182 | 63 | |
AF220121.1 | - | 5 - 6 | 1187 | 63 | ||
AK011082.1 | - | 5 - 6 | 1200 | 63 | ||
BC051632.1 | - | 5 - 6 | 1173 | 6507331 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |