Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009516.3 | NP_033542.2 | 4 - 5 | 1307 | 108 | |
GenBank mRNA | AK011212.1 | - | 4 - 5 | 1083 | 108 | |
AK039572.1 | - | 4 - 5 | 623 | 108 | ||
BC006852.1 | - | 4 - 5 | 1326 | 3156953 | 108 | |
D30743.1 | - | 4 - 5 | 1308 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |