Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285507.1 | NP_001272436.1 | 5 - 6 | 578 | 161 | |
NM_001285512.1 | NP_001272441.1 | 5 - 6 | 605 | 161 | ||
NM_001285513.1 | NP_001272442.1 | 4 - 5 | 369 | 161 | ||
NM_009939.3 | NP_034069.2 | 5 - 6 | 557 | 161 | ||
GenBank mRNA | AF071312.1 | - | 5 - 6 | 467 | 161 | |
AF087688.1 | - | 5 - 6 | 464 | 161 | ||
AK010383.1 | - | 5 - 6 | 504 | 161 | ||
AK037757.1 | - | 5 - 6 | 504 | 161 | ||
AK084421.1 | - | 5 - 6 | 478 | 161 | ||
AK132207.1 | - | 5 - 6 | 474 | 161 | ||
AK132776.1 | - | 5 - 6 | 542 | 161 | ||
AK145225.1 | - | 4 - 5 | 280 | 161 | ||
AK146153.1 | - | 5 - 6 | 468 | 161 | ||
BC023096.1 | - | 5 - 6 | 462 | 3675508 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |