Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010476.3 | NP_034606.3 | 6 - 7 | 821 | 62 | |
GenBank mRNA | AK028380.1 | - | 6 - 7 | 777 | 62 | |
AK050211.1 | - | 6 - 7 | 778 | 62 | ||
AK077914.1 | - | 6 - 7 | 819 | 62 | ||
BC011464.1 | - | 6 - 7 | 785 | 62 | ||
Y15733.1 | - | 6 - 7 | 811 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |