Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026173.3 | NP_080449.1 | 8 - 9 | 1130 | 97 | |
GenBank mRNA | AK004769.2 | - | 8 - 9 | 1058 | 97 | |
AK012926.1 | - | 8 - 9 | 1098 | 97 | ||
AK098149.1 | - | 8 - 9 | 1023 | 97 | ||
AK150656.1 | - | 8 - 9 | 1131 | 97 | ||
BC016527.1 | - | 8 - 9 | 1044 | 3157281 | 97 | |
BC027118.1 | - | 8 - 9 | 1068 | 5359153 | 97 | |
BC037631.1 | - | 8 - 9 | 1087 | 4954391 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |