Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077495.2 | NP_001070963.1 | 7 - 8 | 1538 | 113 | |
GenBank mRNA | BC026146.1 | - | 7 - 8 | 1519 | 3979333 | 113 |
M60651.1 | - | 7 - 8 | 1473 | 113 | ||
U50413.1 | - | 7 - 8 | 1491 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |