Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011420.2 | NP_035550.1 | 2 - 3 | 189 | 130 | |
GenBank mRNA | AK012698.1 | - | 2 - 3 | 159 | 130 | |
AK152495.1 | - | 2 - 3 | 164 | 130 | ||
AK167832.1 | - | 2 - 3 | 187 | 130 | ||
AY821787.1 | - | 2 - 3 | 150 | 130 | ||
BC045158.1 | - | 2 - 3 | 153 | 6486685 | 130 | |
CJ058980.1 | - | 2 - 3 | 190 | 130 | ||
U63294.1 | - | 2 - 3 | 167 | 130 | ||
U77714.1 | - | 2 - 3 | 150 | 130 | ||
Y12835.1 | - | 2 - 3 | 175 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |