Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085376.2 | NP_001078845.1 | 13 - 14 | 4148 | 66 | |
GenBank mRNA | BC094560.1 | - | 3 - 4 | 408 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |