Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NM_001159986.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NM_001159986.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NM_033354.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NM_033354.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NR_027641.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
NR_027641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011742.mm.1 |
TC0100001434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NM_001159986.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NM_001159986.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NM_033354.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NM_033354.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NR_027641.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
NR_027641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011743.mm.1 |
TC0100001434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NM_001159986.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NM_001159986.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NM_033354.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NM_033354.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NR_027641.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
NR_027641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011741.mm.1 |
TC0100001434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NM_001159986.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NM_001159986.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NM_033354.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NM_033354.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NR_027641.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
NR_027641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011739.mm.1 |
TC0100001434.mm.1 |
MTA TC |