Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040399.1 | NP_001035489.1 | 5 - 6 | 495 | 87 | |
GenBank mRNA | AK076732.1 | - | 5 - 6 | 463 | 87 | |
AK160155.1 | - | 5 - 6 | 495 | 87 | ||
BC025528.1 | - | 3 - 4 | 418 | 5324328 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |