Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026876.3 | NP_081152.2 | 10 - 11 | 1723 | 106 | |
GenBank mRNA | AK004493.1 | - | 2 - 3 | 216 | 106 | |
AK146088.1 | - | 10 - 11 | 1724 | 106 | ||
BC022682.1 | - | 5 - 6 | 759 | 106 | ||
BC037478.1 | - | 6 - 7 | 903 | 106 | ||
BC064024.1 | - | 10 - 11 | 1521 | 106 | ||
BC110425.1 | - | 10 - 11 | 1706 | 5369595 | 106 | |
BC132299.1 | - | 10 - 11 | 1581 | 40130576 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |