Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081078.1 | NP_001074547.1 | 13 - 14 | 5004 | 61 | |
GenBank mRNA | AK158042.1 | - | 7 - 8 | 3106 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |