Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173392.4 | NP_775568.1 | 11 - 12 | 3809 | 105 | |
GenBank mRNA | AK122241.1 | - | 11 - 12 | 3790 | 105 | |
BC042669.1 | - | 11 - 12 | 3807 | 4235024 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |