Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028718.2 | NP_082994.1 | 5 - 6 | 1015 | 98 | |
GenBank mRNA | AK014457.1 | - | 2 - 3 | 496 | 98 | |
AY613437.1 | - | 5 - 6 | 1018 | 98 | ||
BC117868.2 | - | 5 - 6 | 989 | 40095476 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |