Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159965.1 | NP_001153437.1 | 5 - 6 | 514 | 81 | |
NM_001159966.1 | NP_001153438.1 | 5 - 6 | 412 | 81 | ||
NM_001159967.1 | NP_001153439.1 | 5 - 6 | 514 | 81 | ||
NM_001159968.1 | NP_001153440.1 | 4 - 5 | 409 | 81 | ||
NM_023884.4 | NP_076373.3 | 5 - 6 | 454 | 81 | ||
NR_027640.1 | 6 - 7 | 619 | 81 | |||
GenBank mRNA | AF312924.1 | - | 4 - 5 | 362 | 81 | |
AK018622.1 | - | 5 - 6 | 456 | 81 | ||
AK019543.1 | - | 4 - 5 | 412 | 81 | ||
AK029049.1 | - | 5 - 6 | 455 | 81 | ||
AK029993.1 | - | 6 - 7 | 592 | 81 | ||
AK033549.1 | - | 5 - 6 | 410 | 81 | ||
AK036803.1 | - | 4 - 5 | 386 | 81 | ||
AK161080.1 | - | 5 - 6 | 481 | 81 | ||
AK165002.1 | - | 5 - 6 | 406 | 81 | ||
AK169368.1 | - | 5 - 6 | 432 | 81 | ||
BC043132.1 | - | 5 - 6 | 419 | 6811441 | 81 | |
BC052663.1 | - | 5 - 6 | 431 | 30051209 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |