Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025456.3 | NP_079732.1 | 4 - 5 | 546 | 60 | |
GenBank mRNA | AK007442.1 | - | 4 - 5 | 501 | 60 | |
AK145027.1 | - | 4 - 5 | 565 | 60 | ||
AK145041.1 | - | 4 - 5 | 565 | 60 | ||
AY882557.1 | - | 4 - 5 | 465 | 60 | ||
BC037622.1 | - | 4 - 5 | 573 | 5373275 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |