Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020557.4 | NP_065582.3 | 1 - 2 | 818 | 53 | |
GenBank mRNA | AK004595.1 | - | 1 - 2 | 653 | 53 | |
AK150512.1 | - | 1 - 2 | 643 | 53 | ||
AK150543.1 | - | 1 - 2 | 645 | 53 | ||
AK150725.1 | - | 1 - 2 | 643 | 53 | ||
AK150885.1 | - | 1 - 2 | 645 | 53 | ||
AK153429.1 | - | 1 - 2 | 647 | 53 | ||
BC027329.1 | - | 1 - 2 | 717 | 3982036 | 53 | |
BC048910.1 | - | 1 - 2 | 54 | |||
BC057565.1 | - | 1 - 2 | 631 | 6401867 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |