Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019432.2 | NP_062305.1 | 1 - 2 | 68 | 93 | |
GenBank mRNA | AK002424.1 | - | 1 - 2 | 87 | 93 | |
AK050134.1 | - | 1 - 2 | 68 | 93 | ||
AK086093.1 | - | 1 - 2 | 73 | 93 | ||
BC024613.1 | - | 1 - 2 | 64 | 4215775 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |